Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020630.1 | NP_001018466.1 | 2 - 3 | 542 | 82 | |
XM_021480399.1 | XP_021336074.1 | 1 - 2 | 418 | 82 | ||
GenBank mRNA | BC095608.1 | - | 2 - 3 | 542 | 82 | |
BC165429.1 | - | 1 - 2 | 245 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |