Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020630.1 | NP_001018466.1 | 5 - 6 | 831 | 73 | |
XM_021480399.1 | XP_021336074.1 | 5 - 6 | 843 | 73 | ||
GenBank mRNA | BC095608.1 | - | 5 - 6 | 831 | 73 | |
BC165429.1 | - | 4 - 5 | 534 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |