Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040307.1 | NP_001035397.1 | 2 - 3 | 531 | 65 | |
XM_021475840.1 | XP_021331515.1 | 2 - 3 | 610 | 65 | ||
GenBank mRNA | BC115288.1 | - | 2 - 3 | 567 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |