Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021470074.1 | XP_021325749.1 | 2 - 3 | 710 | 66 | |
GenBank mRNA | BC115345.1 | - | 3 - 4 | 699 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |