Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fance | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040634.1 | NP_001035724.1 | 2 - 3 | 771 | 98 | |
GenBank mRNA | AY968595.1 | - | 2 - 3 | 771 | 98 | |
BC154582.1 | - | 2 - 3 | 866 | 98 | ||
BC162341.1 | - | 2 - 3 | 794 | 98 | ||
BC162347.1 | - | 2 - 3 | 794 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |