Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn1lab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044895.2 | NP_001038360.2 | 9 - 10 | 1398 | 74 | |
XM_005165770.4 | XP_005165827.1 | 10 - 11 | 2019 | 74 | ||
XM_009302186.3 | XP_009300461.1 | 10 - 11 | 1666 | 74 | ||
XM_021476883.1 | XP_021332558.1 | 9 - 10 | 1345 | 74 | ||
XM_021476884.1 | XP_021332559.1 | 9 - 10 | 1373 | 74 | ||
XM_021476885.1 | XP_021332560.1 | 9 - 10 | 1345 | 74 | ||
XM_021476886.1 | XP_021332561.1 | 9 - 10 | 1345 | 74 | ||
XM_021476887.1 | XP_021332562.1 | 9 - 10 | 1345 | 74 | ||
GenBank mRNA | DQ149504.1 | - | 9 - 10 | 1345 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |