Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045564.2 | NP_001039029.2 | 4 - 5 | 488 | 64 | |
XM_021468901.1 | XP_021324576.1 | 4 - 5 | 486 | 64 | ||
GenBank mRNA | AF155581.1 | - | 4 - 5 | 386 | 64 | |
BC092728.1 | - | 4 - 5 | 398 | 64 | ||
BC135050.1 | - | 4 - 5 | 400 | 64 | ||
BC163781.1 | - | 4 - 5 | 449 | 64 | ||
BC164627.1 | - | 4 - 5 | 426 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |