Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxj1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076706.2 | NP_001070174.2 | 2 - 3 | 775 | 65 | |
GenBank mRNA | EU201184.1 | - | 2 - 3 | 775 | 65 | |
EU599363.1 | - | 1 - 2 | 576 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |