Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077533.1 | NP_001071001.1 | 4 - 5 | 463 | 57 | |
XM_005156072.4 | XP_005156129.1 | 4 - 5 | 497 | 57 | ||
GenBank mRNA | BC125955.1 | - | 4 - 5 | 463 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |