Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc18a3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077550.1 | NP_001071018.1 | 1 | 1747 | 61 | |
GenBank mRNA | BC124354.1 | - | 1 | 1747 | 61 | |
BC165388.1 | - | 1 | 1466 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |