Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mxra8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079960.3 | NP_001073429.3 | 3 - 4 | 450 | 78 | |
GenBank mRNA | BC124787.1 | - | 3 - 4 | 419 | 78 | |
CU638746.1 | - | 3 - 4 | 392 | 78 | ||
EH585789.1 | - | 3 - 4 | 476 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |