Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim62.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080674.1 | NP_001074143.1 | 4 - 5 | 1038 | 83 | |
GenBank mRNA | BC129354.1 | - | 4 - 5 | 1038 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |