Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stxbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089376.1 | NP_001082845.1 | 14 - 15 | 1287 | 73 | |
GenBank mRNA | BC171526.1 | - | 14 - 15 | 1269 | 73 | |
BC171552.1 | - | 14 - 15 | 1269 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |