Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nt5c1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089444.1 | NP_001082913.1 | 1 - 2 | 206 | 83 | |
GenBank mRNA | BC139706.1 | - | 1 - 2 | 206 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |