Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sobpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098618.1 | NP_001092088.1 | 7 - 8 | 763 | 75 | |
GenBank mRNA | DQ655703.1 | - | 7 - 8 | 763 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |