Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casp8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098619.1 | NP_001092089.1 | 2 - 3 | 227 | 92 | |
XM_005165822.4 | XP_005165879.2 | 3 - 4 | 1083 | 92 | ||
GenBank mRNA | DQ812121.1 | - | 2 - 3 | 227 | 92 | |
MG958001.1 | - | 2 - 3 | 227 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |