Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plcd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109700.1 | NP_001103170.1 | 12 - 13 | 2004 | 102 | |
XM_005162701.4 | XP_005162758.1 | 12 - 13 | 2078 | 102 | ||
GenBank mRNA | BC152502.1 | - | 12 - 13 | 2004 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |