Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109703.1 | NP_001103173.1 | 4 - 5 | 653 | 143 | |
XM_009305801.3 | XP_009304076.1 | 4 - 5 | 1056 | 143 | ||
GenBank mRNA | BC151992.1 | - | 4 - 5 | 653 | 143 | |
HM138697.1 | - | 3 - 4 | 561 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |