Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: isl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130962.1 | NP_571037.1 | 4 - 5 | 971 | 85 | |
XM_005165264.4 | XP_005165321.1 | 4 - 5 | 1006 | 85 | ||
XM_021475997.1 | XP_021331672.1 | 4 - 5 | 1012 | 85 | ||
GenBank mRNA | BC060892.1 | - | 4 - 5 | 935 | 85 | |
D21135.1 | - | 4 - 5 | 971 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |