Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jak2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131093.1 | NP_571168.1 | 20 - 21 | 2835 | 73 | |
XM_009295057.3 | XP_009293332.1 | 20 - 21 | 3013 | 73 | ||
GenBank mRNA | AB018222.1 | - | 17 - 18 | 2201 | 73 | |
AJ005690.1 | - | 20 - 21 | 2835 | 73 | ||
BC162596.1 | - | 20 - 21 | 2794 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |