Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoxc4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131122.2 | NP_571197.1 | 1 - 2 | 443 | 70 | |
GenBank mRNA | BC092923.1 | - | 1 - 2 | 443 | 70 | |
BC164612.1 | - | 1 - 2 | 469 | 70 | ||
DQ060552.1 | - | 1 - 2 | 218 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |