Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131234.2 | NP_571309.2 | 3 - 4 | 794 | 61 | |
GenBank mRNA | BC124483.1 | - | 3 - 4 | 722 | 61 | |
BC164788.1 | - | 2 - 3 | 730 | 61 | ||
U18311.1 | - | 3 - 4 | 796 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |