Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pla2g4aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131295.2 | NP_571370.1 | 14 - 15 | 1657 | 63 | |
GenBank mRNA | BC162082.1 | - | 13 - 14 | 1585 | 63 | |
U10330.1 | - | 14 - 15 | 1658 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |