Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131506.2 | NP_571581.2 | 3 - 4 | 702 | 133 | |
XM_021466746.1 | XP_021322421.1 | 3 - 4 | 892 | 133 | ||
GenBank mRNA | AF239664.1 | - | 3 - 4 | 704 | 133 | |
AF253325.1 | - | 3 - 4 | 717 | 133 | ||
BC163475.1 | - | 3 - 4 | 693 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |