Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131881.1 | NP_571956.1 | 1 - 2 | 229 | 62 | |
GenBank mRNA | AF398433.1 | - | 1 - 2 | 229 | 62 | |
AF414133.1 | - | 1 - 2 | 232 | 62 | ||
BC095012.1 | - | 1 - 2 | 238 | 62 | ||
BC164337.1 | - | 1 - 2 | 239 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |