Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153651.2 | NP_705937.2 | 3 - 4 | 1318 | 110 | |
GenBank mRNA | AF398518.1 | - | 3 - 4 | 1365 | 110 | |
AY045753.1 | - | 3 - 4 | 1328 | 110 | ||
BC128814.1 | - | 3 - 4 | 1318 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |