Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thbs4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173226.1 | NP_775333.1 | 16 - 17 | 2198 | 71 | |
GenBank mRNA | AY081009.1 | - | 16 - 17 | 2198 | 71 | |
BC095593.1 | - | 16 - 17 | 2208 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |