Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anxa11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181765.2 | NP_861430.2 | 9 - 10 | 1170 | 91 | |
NM_183410.2 | NP_899670.1 | 9 - 10 | 1041 | 91 | ||
GenBank mRNA | AY178801.1 | - | 9 - 10 | 1162 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |