Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183339.1 | NP_899182.1 | 5 - 6 | 1258 | 58 | |
XM_005168611.4 | XP_005168668.1 | 6 - 7 | 1687 | 58 | ||
XM_009301230.3 | XP_009299505.1 | 6 - 7 | 1634 | 58 | ||
GenBank mRNA | AY151388.1 | - | 5 - 6 | 1258 | 58 | |
AY197606.1 | - | 5 - 6 | 898 | 58 | ||
AY277631.1 | - | 5 - 6 | 1028 | 58 | ||
AY294284.1 | - | 5 - 6 | 1258 | 58 | ||
BC115243.1 | - | 5 - 6 | 1252 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |