Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rprd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199556.1 | NP_955850.1 | 2 - 3 | 369 | 84 | |
XM_005162418.3 | XP_005162475.1 | 2 - 3 | 460 | 84 | ||
XM_005162422.1 | XP_005162479.1 | 2 - 3 | 434 | 84 | ||
XM_005162423.1 | XP_005162480.1 | 2 - 3 | 434 | 84 | ||
GenBank mRNA | BC048885.1 | - | 2 - 3 | 369 | 84 | |
BC068349.1 | - | 2 - 3 | 380 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |