Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rprd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199556.1 | NP_955850.1 | 7 - 8 | 1067 | 64 | |
XM_005162418.3 | XP_005162475.1 | 7 - 8 | 1161 | 64 | ||
XM_005162422.1 | XP_005162479.1 | 7 - 8 | 1120 | 64 | ||
XM_005162423.1 | XP_005162480.1 | 7 - 8 | 1117 | 64 | ||
GenBank mRNA | BC048885.1 | - | 7 - 8 | 1067 | 64 | |
BC068349.1 | - | 7 - 8 | 1078 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |