Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc43a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327986.1 | NP_001314915.1 | 3 - 4 | 116 | ||
GenBank mRNA | BC044509.1 | - | 3 - 4 | 608 | 116 | |
BC164469.1 | - | 2 - 3 | 365 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |