Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199902.1 | NP_956196.1 | 6 - 7 | 1329 | 96 | |
GenBank mRNA | BC054664.1 | - | 6 - 7 | 1329 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |