Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199909.2 | NP_956203.1 | 10 - 11 | 1142 | 83 | |
GenBank mRNA | AY162326.1 | - | 10 - 11 | 1142 | 83 | |
BC053124.1 | - | 10 - 11 | 1123 | 83 | ||
BC150356.1 | - | 10 - 11 | 1146 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |