Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: seh1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199923.2 | NP_956217.2 | 4 - 5 | 744 | 69 | |
XM_021468000.1 | XP_021323675.1 | 4 - 5 | 739 | 69 | ||
GenBank mRNA | AY398414.1 | - | 4 - 5 | 744 | 69 | |
BC051159.1 | - | 4 - 5 | 70 | |||
CT658356.2 | - | 4 - 5 | 618 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |