Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asnsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200095.2 | NP_956389.2 | 5 - 6 | 1944 | 124 | |
GenBank mRNA | BC051619.1 | - | 3 - 4 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |