Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2p6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200139.1 | NP_956433.1 | 1 - 2 | 300 | 99 | |
GenBank mRNA | BC044463.1 | - | 1 - 2 | 300 | 99 | |
BC165616.1 | - | 1 - 2 | 224 | 99 | ||
DQ478813.1 | - | 1 - 2 | 214 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |