Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200595.1 | NP_956889.1 | 1 - 2 | 492 | 70 | |
GenBank mRNA | BC056756.1 | - | 1 - 2 | 492 | 70 | |
BC066397.1 | - | 1 - 2 | 431 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |