Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: waslb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201059.2 | NP_957353.2 | 5 - 6 | 647 | 99 | |
XM_021474971.1 | XP_021330646.1 | 5 - 6 | 423 | 99 | ||
GenBank mRNA | BC044180.1 | - | 5 - 6 | 697 | 99 | |
BC066424.1 | - | 5 - 6 | 647 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |