Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arrb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201124.1 | NP_957418.1 | 12 - 13 | 1139 | 131 | |
XM_009303039.3 | XP_009301314.1 | 11 - 12 | 1224 | 131 | ||
XM_009303040.3 | XP_009301315.1 | 11 - 12 | 1235 | 131 | ||
GenBank mRNA | BC054595.1 | - | 12 - 13 | 1139 | 131 | |
BC164508.1 | - | 12 - 13 | 1034 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |