Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: e2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197300.1 | NP_001184229.1 | 2 - 3 | 421 | 119 | |
NM_205615.2 | NP_991178.1 | 2 - 3 | 421 | 119 | ||
GenBank mRNA | AY398338.1 | - | 2 - 3 | 366 | 119 | |
BC098598.1 | - | 2 - 3 | 417 | 119 | ||
EB911484.1 | - | 2 - 3 | 297 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |