Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dao.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205694.1 | NP_991257.1 | 7 - 8 | 658 | 90 | |
XM_021476050.1 | XP_021331725.1 | 6 - 7 | 617 | 90 | ||
GenBank mRNA | BC065882.1 | - | 7 - 8 | 658 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |