Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ehhadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207068.1 | NP_996951.1 | 2 - 3 | 204 | 89 | |
GenBank mRNA | BC066545.1 | - | 2 - 3 | 204 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |