Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212596.1 | NP_997761.1 | 2 - 3 | 283 | 67 | |
XM_005159648.3 | XP_005159705.1 | 2 - 3 | 67 | |||
GenBank mRNA | BC053160.1 | - | 2 - 3 | 283 | 67 | |
KJ651951.1 | - | 2 - 3 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |