Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213066.1 | NP_998231.1 | 3 - 4 | 472 | 108 | |
GenBank mRNA | BC045496.1 | - | 3 - 4 | 472 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |