Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thap12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213081.1 | NP_998246.1 | 4 - 5 | 383 | 71 | |
GenBank mRNA | BC044456.1 | - | 4 - 5 | 383 | 71 | |
BC165660.1 | - | 4 - 5 | 360 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |