Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: laptm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214685.2 | NP_999850.2 | 4 - 5 | 500 | 66 | |
GenBank mRNA | BC058878.1 | - | 4 - 5 | 527 | 66 | |
BC155748.1 | - | 4 - 5 | 500 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |