Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: th2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001829.1 | NP_001001829.1 | 3 - 4 | 563 | 127 | |
XM_005164734.4 | XP_005164791.1 | 3 - 4 | 891 | 127 | ||
GenBank mRNA | AY620823.1 | - | 3 - 4 | 563 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |