Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: puf60b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002121.1 | NP_001002121.1 | 9 - 10 | 1060 | 113 | |
XM_009303395.3 | XP_009301670.1 | 10 - 11 | 1249 | 113 | ||
GenBank mRNA | BC071467.1 | - | 9 - 10 | 1060 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |