Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002566.1 | NP_001002566.1 | 1 - 2 | 585 | 110 | |
GenBank mRNA | BC076234.1 | - | 1 - 2 | 585 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |